missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Centrifree™ Filters
Description
The Centrifree™ Ultrafiltration device with Ultracel regenerated cellulose membrane is ideal for separating free from bound microsolutes in serum, plasma, and other biological samples. The device is licensed for research use and in vitro diagnostic use. The device holds a sample volume of 1 mL with a hold-up volume of 10 μL.
Features and Benefits:
- Separate unbound (free) therapeutic drugs, testosterone, thyroxin, etc.
- Enables accuracy in binding studies
- Critical for new drug investigations
- Facilitates deproteinization to reduce complexity or matrix interference of biological samples
- For research and in vitro diagnostic use
Specifications
Specifications
| Filter Type | Centrifuge Filter |
| Format | Microcentrifuge, Spin Column |
| For Use With (Equipment) | Centrifuge |
| Material | Polystyrene, Polyethylene, SAN |
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 30 kDa |
| Sample Volume | 1 mL |
| Sterility | Non-sterile |
| Type | Centrifugal Filter |
| Volume (Metric) | 1 mL |