missing translation for 'onlineSavingsMsg'
Learn More

Welch™ Centering Ring Rubber O-ring

Catalog No. 01184162
Change view
Click to view available options
For Use With (Equipment):
Stainless steel centering ring
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01184162 Stainless steel centering ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01184162 Supplier Welch™ Supplier No. 304801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with stainless steel centering ring

  • For use with stainless steel centering ring
  • Replaceable fluoroelastomer 0-ring seal

Specifications

For Use With (Equipment) Stainless steel centering ring
Product Type Rubber O-ring
Material Rubber
Color Black