Learn More
Mopec Center Approach Wall Mount Autopsy Sink
Save time and space with wall mounted autopsy dissection sink. Mopec™ Center Approach Wall Mount Autopsy Sink features a cart tray that hangs over the sink to allow for proper fluid drainage.
Supplier:  Mopec MPCCC100
Catalog No. 22444015
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Stainless steel construction
  • Center sink approach
  • Hydro aspirator with reversing flow valve
  • Includes hose hanger and 8 foot clear PVC tubing
  • Two cold water body rinse valve with vacuum breaker and serrated hose fitting
  • Two hot and cold water fixture with wrist blade handles
  • 8 inch swing spout with vacuum breaker
  • Four interchangeable removable stainless steel grid plates
  • Grid plates constructed with .5 in. recess and .375 in. diameter holes on 2 in. square centers
  • Two GFCI duplex outlets with waterproof covers
  • Two instrument drawers – Flush to the table when closed and fitted
  • Flush inset handles at the top to keep fluid from accumulating in the drawers
  • Two hand spray assemblies
  • Heavy duty brass chrome plated hand piece
  • J hook to hold hand piece
  • Concealed pressure control shut off
Warranty
1 Year Warranty
Specifications
Specifications
Autopsy Sink
28 in.
114 in.
154.94 cm
Stainless Steel
71.12 cm
61 in.
289.56 cm
Product Suggestions
Product Suggestions