Learn More
Abcam Inc CENTA, beta-lactamase substrate, Yellow chromogenic beta-lactamase substrate, 10MG
Catalog No. 502705098
Encompass
MW 535.6 Da, Purity >98%. Yellow chromogenic -lactamase substrate. Hydrolyses readily in the presence of all -lactamases except for Aeromonas hydrophila metalloenzyme. Suitable for kinetic studies, the detection of the enzymes in crude extracts and chromatographic fractions. Absorbs light at 346 nm ( = -2500 M- cm-) or 405 nm ( = +6400 M- cm-).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502705098 Supplier Abcam Inc Supplier No. AB14562610MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 535.6 Da, Purity >98%. Yellow chromogenic -lactamase substrate. Hydrolyses readily in the presence of all -lactamases except for Aeromonas hydrophila metalloenzyme. Suitable for kinetic studies, the detection of the enzymes in crude extracts and chromatographic fractions. Absorbs light at 346 nm ( = -2500 M- cm-) or 405 nm ( = +6400 M- cm-).
The product is subject to the following: Abcam Restricted Use Statement