missing translation for 'onlineSavingsMsg'
Learn More

Foxx Life Sciences EZBio MTO Centrifuge Tube Assembly, 15mL, Polypropylene (PP) Centrifuge Tube, Sampling Tube, Single Use Technology with Pureweld Tubing and Pinchpipe, Vented with Diptube, CPC MPC, Sterile, 10/CS
GREENER_CHOICE

Catalog No. 503029732
Encompass

The sterile EZBio Centrifuge Tube Assembly is a single use closed loop system designed for aseptic sampling and sterile transfer.
All components are USP Class VI resin, assembled in a Class 7 cleanroom, and gamma-irradiated in the USA. These assemblies feature a closed cap, an 18 inlet line (1/8 x 1/4 TPE tubing) with a pinch clamp, pinch pipe, plug or luer or CPC MPC/AQS, a 1.5 vent line with PTFE hydrophobic filter, and a dip tube. A centrifuge tube stand securely holds either size. The polypropylene resin meets FDA and USP standards.

Features
Designed for contamination-free sampling/transfer within closed systems
Leak-proof seal with a platinum-cured silicone gasket
Available in 15mL and 50mL with six TPE tubing options and four fitting types.
Withstands up to 4 psi; pinch pipe enables quick aseptic separation

Applications
Cell Harvest & Downstream Processing
Drug Substance Purification
Drug Product Formulation
Buffer & Media Preparation

Catalog No. 503029732 Supplier Foxx Life Sciences Supplier No. 58540D1FLS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The sterile EZBio Centrifuge Tube Assembly is a single use closed loop system designed for aseptic sampling and sterile transfer.
All components are USP Class VI resin, assembled in a Class 7 cleanroom, and gamma-irradiated in the USA. These assemblies feature a closed cap, an 18 inlet line (1/8 x 1/4 TPE tubing) with a pinch clamp, pinch pipe, plug or luer or CPC MPC/AQS, a 1.5 vent line with PTFE hydrophobic filter, and a dip tube. A centrifuge tube stand securely holds either size. The polypropylene resin meets FDA and USP standards.

Features
Designed for contamination-free sampling/transfer within closed systems
Leak-proof seal with a platinum-cured silicone gasket
Available in 15mL and 50mL with six TPE tubing options and four fitting types.
Withstands up to 4 psi; pinch pipe enables quick aseptic separation

Applications
Cell Harvest & Downstream Processing
Drug Substance Purification
Drug Product Formulation
Buffer & Media Preparation