missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cellulose triacetate

Catalog No. p-84143
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC177822500 250 g Plastic bottle
AC177825000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC177822500 Supplier Thermo Scientific Chemicals Supplier No. 177822500
Only null left

CAS: 9012-09-3 | (C24H32O16)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9012-09-3
Molecular Formula (C24H32O16)n
MDL Number MFCD00132680
InChI Key NNLVGZFZQQXQNW-ADJNRHBOSA-N
Synonym triacetylcellulose, cellulose triacetate, beta-cellotriose undecaacetate, 2r,3r,4s,5r,6s-4,5-bis acetyloxy-6-2r,3r,4s,5r,6s-4,5,6-tris acetyloxy-2-acetyloxy methyl oxan-3-yl oxy-3-2s,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-acetyloxy methyl oxan-2-yl oxy oxan-2-yl methyl acetate
PubChem CID 44263853
IUPAC Name [(2R,3R,4S,5R,6S)-4,5-diacetyloxy-3-[(2S,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxy-6-[(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-2-(acetyloxymethyl)oxan-3-yl]oxyoxan-2-yl]methyl acetate

Specifications

Color White to Yellow
Physical Form Pellets or Rods
Merck Index 10, 1926
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform, methylene dichloride and, tetrachloroethane
Quantity 250 g
SMILES CC(=O)OCC1C(C(C(C(O1)OC2C(OC(C(C2OC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC3C(C(C(C(O3)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Cellulose triacetate

RUO – Research Use Only