missing translation for 'onlineSavingsMsg'
Learn More

Cellulose powder, microcrystallline, >97%, MP Biomedicals™

Catalog No. p-4405598
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19149983 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19149983 Supplier MP Biomedicals Inc Supplier No. 0219149983
Only null left

Chemical Identifiers

CAS 9004-34-6
Molecular Formula (C6H10O5)n

Specifications

Color White
Physical Form Powder
Assay Percent Range >97%
Quantity 250 g
Chemical Name or Material Cellulose powder