missing translation for 'onlineSavingsMsg'
Learn More

Cellulose powder, For column chromatography, MilliporeSigma™ Supelco™

Catalog No. 111015798
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015798 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015798 Supplier Merck Emd Millipore Supplier No. 221831KGF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00081512
Synonym Cellulose powder; Cotton linters

Specifications

Physical Form Powder
Quantity 1 kg
Grade Column Chromatography
RTECSNumber FJ5691460