missing translation for 'onlineSavingsMsg'
Learn More

Cellulose microcrystalline, extra pure, average particle size 90 μm

Catalog No. ac382310250
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC382310250 25 kg Fiber Drum
AC382312500 250 g Glass bottle
AC382310010 1 kg Glass bottle
AC382310050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC382310250 Supplier Thermo Scientific Chemicals Supplier No. 382310250
Only null left

CAS: 9004-34-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid

Chemical Identifiers

CAS 9004-34-6

Specifications

Color White
pH 5 to 7
Physical Form Fine Crystalline Powder or Granular Powder
Linear Formula (C12H20O10)n
Solubility Information Solubility in water: insoluble.
Quantity 25 kg
Grade Extra Pure
Infrared Spectrum Authentic
Loss on Drying 1.5% max.
Packaging Fiber Drum
Particle Size 250 Micron <1% min., 75 Micron >45% min., 32 Micron >70% min.
Residue after Ignition 0.05% max.
Chemical Name or Material Cellulose microcrystalline
Show More Show Less

RUO – Research Use Only