Learn More
Cellulose acetate phthalate , MilliporeSigma™ Supelco™
Cellulose acetate phthalate (CAP) is a microbicide which inactivates sexually transmitted disease (STD) pathogens like HIV-1. CAP has been used for enteric film coating of tablets and capsules.
$184.74 - $741.19
Chemical Identifiers
MDL Number MFCD00072769
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111015801
View Documents
Merck Emd Millipore
22192100GF
100 g
Each for $184.74
Only null left
111015800
View Documents
Merck Emd Millipore
22192500GF
500 g
Each for $741.19
Only null left
Description
Description
Specifications
Chemical Identifiers
MFCD00072769
Specifications
9004-38-0
C116H116O64
100 g
Poly Bottle
MFCD00072769
2534.12
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'