missing translation for 'onlineSavingsMsg'
Learn More

Cellulose Acetate Butyrate, Butyryl Content 35-39%

Catalog No. ac404255000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC404255000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC404255000 Supplier Thermo Scientific Chemicals Supplier No. 404255000
Only null left

CAS: 9004-36-8 | (C12H14O10)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-36-8
Molecular Formula (C12H14O10)n
MDL Number MFCD00081513

Specifications

Color White
Physical Form Crystalline Powder
Melting Point 127.0°C to 240.0°C
Avg. Mol. Wt. or Mol. Wt. Range 70000 (Mn)
Solubility Information Solubility in water: negligible. Other solubilities: soluble in ketones, organic acetates, lactates,, methylene, ethylene, propylene chlorides and, high-boiling solvents
Quantity 500 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Refractive Index 1.475
Viscosity 17 to 28 (sec.)
Chemical Name or Material Cellulose acetate butyrate, Butyryl Content 35-39%
Show More Show Less

RUO – Research Use Only