Learn More
Celltreat Scientific Products 150mm x 20mm Tissue Culture Treated Dish, Sterile
Supplier: Celltreat Scientific Products 229650
Cell Culture Dishes have thicker and more durable plastic than Petri Dishes. Cell culture treatment process provides consistent and even hydrophilic surface for improved cell attachment. Manufactured from medical grade virgin polystyrene. 35, 60, 70 and 100mm dishes available with a gripping feature for improved handling. Vented lids contain molded spacers to allow gas exchange, and stacking rings for increased stability.