missing translation for 'onlineSavingsMsg'
Learn More

Promega CellTiter-Glo™ One Solution Assay

Catalog No. PRG8462
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG8462 500 mL
PRG8461 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG8462 Supplier Promega Supplier No. G8462

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Luminescent assay measures number of viable cells in culture.

  • Luminescent Assay Measures Number of Viable Cells in Culture
  • Frozen, ready-to-use assay eliminates several protocol steps

Specifications

Quantity 500 mL