missing translation for 'onlineSavingsMsg'
Learn More

Promega CellTiter-Glo™ 3D Cell Viability Assay

Catalog No. PRG9683
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9683 100 mL
PRG9681 10 mL
PRG9682 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG9683 Supplier Promega Supplier No. G9683

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The CellTiter-Glo™ 3D Cell Viability Assay is a homogeneous method to determine the number of viable cells in 3D cell culture based on quantitation of the ATP present, which is a marker for the presence of metabolically active cells.

  • A Cell Viability Assay Validated for 3D Microtissue Cultures
  • Accurate 3D cytotoxicity determination
  • Easy assay implementation
  • Simple, 30-minute protocol

Specifications

Quantity 100 mL
Content And Storage -30°C to -10°C