missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLSTAR Suspension Culture Flasks
GREENER_CHOICE

Catalog No. 07000279
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
Closure Type:
Filter
Screw
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Type
07000279 250 mL Screw
07000227 50 mL Filter
07000231 50 mL Screw
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07000279 Supplier Greiner Bio-One Supplier No. 658190
Only null left

Improved cell adhesion through physical surface treatment. Greiner Bio-One CELLSTAR Suspension Cell Culture Flasks is free of detectable DNase, RNase and human DNA.

  • Hydrophobic surface, ideal for suspension cultures and hybridoma
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Graduation on the side
  • Canted neck style
  • TC surface treatment: Suspension

Specifications

Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens; Contents non-cytotoxic
Closure Color White
Closure Material HDPE
Capacity (Metric) 250 mL
Material Polystyrene
Neck Style Canted
No. per Case 120
No. per Pack 5
Graduated Yes
Sterility Sterile
Closure Type Screw
Show More Show Less