Learn More
Greiner Bio-One CELLSTAR Small Volume™ 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Color:White, Clear Bottom; Sterility:Non-sterile; Lid:Without Lid
Supplier:  Greiner Bio-One 788095
Catalog No. 07-000-034
Only null left
Explore available promotions
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Compatible with automated systems
  • Stackable
  • Alphanumeric well coding
  • Growth area per well: 2.7mm²
  • Working volume per well:
Specifications
Specifications
No
Without Lid
Medium Binding
Polystyrene
Non-sterile
28 μL
80
Clear
Standard
16 x 24
White, Clear Bottom
384
Cell Culture
Flat Bottom (F-well)
10
Product Suggestions
Product Suggestions