missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLSTAR μClear™ 96-well, Cell Repellent-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

Supplier:  Greiner Bio-One 655976SIN

Catalog No. 07001004


Only null left

Description

Description

  • Does not degrade or leach under common cell culture conditions
  • Effectively inhibits cell adhesion

Applications:

  • Spheroid cultures and stem cell aggregate formation
  • Suspension culture of semi-adher
Specifications

Specifications

Standard
8 x 12
Polystyrene
Sterile
Flat Bottom (F-well)
With Lid
Black, Clear Bottom
96
Cell Repellent
Cell culture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions