missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLSTAR 384 Well Cell Repellent Microplate with Lid

Catalog No. 07000899
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07000899 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07000899 Supplier Greiner Bio-One Supplier No. 781970

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains modified cell-repellent surface.

  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Non-cytotoxic

Specifications

Format Standard
Lid With Lid
Array 16 x 24
Autoclavable Not Autoclavable
Color Clear
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Sterile
Total Volume (Metric) Well 131 μL
Well Shape Rounded Square
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 60
No. per Pack 1
Show More Show Less