missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Color: White; Sterility: Sterile; Lid: With Lid
$866.11 - $3,380.43
Specifications
| Format | Standard |
|---|---|
| Lid | With Lid |
| Array | 16 x 24 |
| Color | White |
| Material | Polystyrene |
Description
- Made from polystyrene
- Tissue-culture treated
- Solid or μClear™ flat bottom
- With lid
Specifications
| Standard | |
| 16 x 24 | |
| Polystyrene | |
| Sterile |
| With Lid | |
| White | |
| 384 | |
| Cell Culture |