Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid
Supplier:  Greiner Bio-One 781090
 View more versions of this product
Catalog No. 07000062
Only null left
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Non-cytotoxicCompatible with automated systemsFlexible working volume
  • Rounded square well profile
  • Stackable
  • Alphanumeric well cod
Specifications
Chemical Identifiers
Standard
16 x 24
Polystyrene
15 to 110 μL
Cell Culture
Rounded Square
With Lid
Black, Clear Bottom
384
Sterile
131 μL
Specifications
Standard
16 x 24
Black, Clear Bottom
384
Sterile
131 μL
120
With Lid
20/Pk., 120/Cs.
Polystyrene
15 to 110 μL
Cell Culture
Rounded Square
20
Product Suggestions
Product Suggestions