Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Color:White, Clear Bottom; Sterility:Sterile; Lid:With Lid
Supplier:  Greiner Bio-One 781098
Catalog No. 07000076
Only null left
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Non-cytotoxicCompatible with automated systemsFlexible working volume
  • Rounded square well profile
  • Stackable
  • Alphanumeric well cod
Specifications
Specifications
Standard
16 x 24
White, Clear Bottom
Polystyrene
15 to 110 μL
Cell Culture
Rounded Square
8
With Lid
8/Pk., 32/Cs.
Clear Bottom
384
Sterile
131 μL
32
32/Cs.
Product Suggestions
Product Suggestions