Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Color:White; Sterility:Sterile; Lid:Without Lid
Supplier: Greiner Bio-One 781073
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic Non-cytotoxicCompatible with automated systemsFlexible working volumeRounded square well profile
- Stackable
- Alphanumeric well cod
Specifications
| Standard | |
| 16 x 24 | |
| Polystyrene | |
| 15 to 110 μL | |
| Cell Culture | |
| Rounded Square | |
| 10 |
| Without Lid | |
| White | |
| 384 | |
| Sterile | |
| 131 μL | |
| 40 | |
| 40/Cs. |