missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate

Catalog No. 07001038
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07001038 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07001038 Supplier Greiner Bio-One Supplier No. 78109099

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Black Lid

  • Made from polystyrene
  • Tissue-culture treated
  • Solid or μClear™ flat bottom
  • With lid

Specifications

Format Standard
Lid With Black Lid
Array 16 x 24
Packaging 20/Pk., 120/Cs.
Color Black, Clear Bottom
Product Type Cell Culture Plate
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
Show More Show Less