Learn More
Alkali Scientific CellPro™ Spectrophotometer Cuvette, PS, 10mm light path, 1000/Pk, 1.5mL semi-micro, square opening
Catalog No. 503027598
Encompass
Our disposable cuvettes are compatible with most popular spectrophotometers. Manufactured from high quality, optically clear polystyrene, they are intended for determinations in the 340-800nm (visible) wavelength range. Cuvettes are available in three sizes: 1.5mL micro, 2.5mL semi-micro and 4.5mL macro. All sizes are 12.5mm square, 45mm height and are compatible with various spectrophotometer models including: Carolina, Spectronic200, Spectronic20 and 20D, SpectronicEducator, Genesys™ and Accuris SmartDrop™. Fits most spectrophotometers and photometers Polystyrene for use in visible range (340-800nm) Protective packaging dust-proof and non-scratch Absorption reproducibility error of less than 1%**Caps are available and can be ordered separately.
Catalog No. 503027598 Supplier Alkali Scientific Supplier No. EP4103
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Our disposable cuvettes are compatible with most popular spectrophotometers. Manufactured from high quality, optically clear polystyrene, they are intended for determinations in the 340-800nm (visible) wavelength range. Cuvettes are available in three sizes: 1.5mL micro, 2.5mL semi-micro and 4.5mL macro. All sizes are 12.5mm square, 45mm height and are compatible with various spectrophotometer models including: Carolina, Spectronic200, Spectronic20 and 20D, SpectronicEducator, Genesys™ and Accuris SmartDrop™. Fits most spectrophotometers and photometers Polystyrene for use in visible range (340-800nm) Protective packaging dust-proof and non-scratch Absorption reproducibility error of less than 1%**Caps are available and can be ordered separately.