Learn More
Alkali Scientific CellPro™ Serological Pipets, Individually Wrapped, Sterile, 1mL, Bagged, 400/Case
Catalog No. 503028251
Encompass
CellPro serological pipettes, or volumetric transfer pipettes, are a common laboratory instrument which is vital for all laboratory operations involving transferring volumes of liquid of from 1mL to 50mL. They are made from 100% polystyrene and featuere bi-directional graduations along with a generous amount of negative graduations. Individually wrapped Gamma Sterilized Color coded paper/plastic wrapping for pop-through or peel-apart techniques Stretched, Non-welded Design Bi-directional and extra Negative Graduation Marks Filter Plug 100% Polystyrene construction RNase/DNase, Pyrogen and Cytotoxin Free
Catalog No. 503028251 Supplier Alkali Scientific Supplier No. SP201BCS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CellPro serological pipettes, or volumetric transfer pipettes, are a common laboratory instrument which is vital for all laboratory operations involving transferring volumes of liquid of from 1mL to 50mL. They are made from 100% polystyrene and featuere bi-directional graduations along with a generous amount of negative graduations. Individually wrapped Gamma Sterilized Color coded paper/plastic wrapping for pop-through or peel-apart techniques Stretched, Non-welded Design Bi-directional and extra Negative Graduation Marks Filter Plug 100% Polystyrene construction RNase/DNase, Pyrogen and Cytotoxin Free