Learn More
Alkali Scientific CellPro™ Penicillin, Streptomycin, and Glutamine Solution, 100ml, 10/Case
Catalog No. 503026981
Encompass
Effective against gram negative and gram positive microorganisms. Penicillin interferes with the final stage of synthesis of the bacterial cell wall. Streptomycin inhibits elongation at transpeptidation step during protein synthesis and binds to 30S subunit, causing misreading. Common working concentration for penicillin is 100 U/mL; for Streptomycin: 100 g/mL. Penicillin/Streptomycin is a broad spectrum bacteriostatic with activity against gram negative and gram-positive organisms. ****This item is required to ship on dry ice. Overnight Shipping required, For Research Use Only (RUO)
Catalog No. 503026981 Supplier Alkali Scientific Supplier No. FB83CS
Effective against gram negative and gram positive microorganisms. Penicillin interferes with the final stage of synthesis of the bacterial cell wall. Streptomycin inhibits elongation at transpeptidation step during protein synthesis and binds to 30S subunit, causing misreading. Common working concentration for penicillin is 100 U/mL; for Streptomycin: 100 g/mL. Penicillin/Streptomycin is a broad spectrum bacteriostatic with activity against gram negative and gram-positive organisms. ****This item is required to ship on dry ice. Overnight Shipping required, For Research Use Only (RUO)