Learn More
Alkali Scientific CellPro™ Molecular Biological Grade Water, 500mL, 10/Case
Catalog No. 503026627
Encompass
CellPro™ Molecular Biological Grade Water is suitable for use in molecular biology applications which demand a high quality of water and assurance that the water is free fromDNaseandRNasecontamination. Molecular Grade Water can be used for making reagents and rinsing glass/plastic wares for DNA and RNA research.
Catalog No. 503026627 Supplier Alkali Scientific Supplier No. DC42311CS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CellPro™ Molecular Biological Grade Water is suitable for use in molecular biology applications which demand a high quality of water and assurance that the water is free fromDNaseandRNasecontamination. Molecular Grade Water can be used for making reagents and rinsing glass/plastic wares for DNA and RNA research.