Learn More
Alkali Scientific CellPro™ HBSS without calcium, magnesium, or phenol red, 500ml
Catalog No. 503028735
Encompass
Balanced salt solution for cell culture applications (liquid form). All formulations incorporate a rigorous specification and approval process. Products with the LS and FS extension are supplied in convenient HyQPAK six-packs. Buffer cell culture growth with CellPro™ 1X Balanced Salt Solutions, available in lot sizes that facilitate scale-up and manufacturing.
Catalog No. 503028735 Supplier Alkali Scientific Supplier No. HBSS02
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Balanced salt solution for cell culture applications (liquid form). All formulations incorporate a rigorous specification and approval process. Products with the LS and FS extension are supplied in convenient HyQPAK six-packs. Buffer cell culture growth with CellPro™ 1X Balanced Salt Solutions, available in lot sizes that facilitate scale-up and manufacturing.