Learn More
Alkali Scientific CellPro™ Ham's Nutrient Mixture F12, with L-glutamine, 500ml
Catalog No. 503027599
Encompass
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Catalog No. 503027599 Supplier Alkali Scientific Supplier No. F12002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.