Learn More
Alkali Scientific CellPro™ DMEM/F12 1:1 medium, with HEPES; without L-glutamine, 500ml
Catalog No. 503028602
Encompass
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Catalog No. 503028602 Supplier Alkali Scientific Supplier No. F12005
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.