Learn More
Alkali Scientific CellPro™ DMEM/F12 1:1 medium, with L-glutamine; without HEPES, 500 ml, 10/Case
Catalog No. 503028734
Encompass
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Catalog No. 503028734 Supplier Alkali Scientific Supplier No. F12004CS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.