Learn More
Alkali Scientific CellPro™ DEPC treated Ultra Pure Water, 1L, 10/case
Catalog No. 503027123
Encompass
DEPC Treated Water (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded byRNases. Water is usually treated with 0.1% v/v DEPC for at least 2 hours at 37 C and then autoclaved (at least 15 min) to inactivate traces of DEPC. Inactivation of DEPC in this manner yields CO 2 and ethanol.
Catalog No. 503027123 Supplier Alkali Scientific Supplier No. DC42001CS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
DEPC Treated Water (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded byRNases. Water is usually treated with 0.1% v/v DEPC for at least 2 hours at 37 C and then autoclaved (at least 15 min) to inactivate traces of DEPC. Inactivation of DEPC in this manner yields CO 2 and ethanol.