Learn More
Alkali Scientific CellPro™ Deep Well Plate, 96 Round Well, 1.2mL, 42.5mm height, F-bottom, for ABgene™ , Black Alphanumeric Printing, PP, 10 bags of 5 pieces, 50 / CS, Sterile
Catalog No. 503029461
Encompass
The CellPro™ line of Deep Well Plates is compatible with all popular automated research platforms and robotic equipment. These plates are useful for bacterial cultures, centrifugation (4000xg max RCF) and general sample storage. They can be used with silicone sealing mats and various types of sealing film. All plates are produced in a class 100,000 cleanroom facility and are ANSI / SLAS 1-2004 compliant. Although all OptiWell™ plates are autoclavable, several types are also available presterilized by gamma irradiation in accordance with ISO 13485. All plates are certified RNase/DNase free and non-pyrogenic. For automated research platforms / robotic equipment Minimal binding / liquid retention Black printed alpha-numerics Virgin, medical-grade polypropylene
Catalog No. 503029461 Supplier Alkali Scientific Supplier No. MPD49S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The CellPro™ line of Deep Well Plates is compatible with all popular automated research platforms and robotic equipment. These plates are useful for bacterial cultures, centrifugation (4000xg max RCF) and general sample storage. They can be used with silicone sealing mats and various types of sealing film. All plates are produced in a class 100,000 cleanroom facility and are ANSI / SLAS 1-2004 compliant. Although all OptiWell™ plates are autoclavable, several types are also available presterilized by gamma irradiation in accordance with ISO 13485. All plates are certified RNase/DNase free and non-pyrogenic. For automated research platforms / robotic equipment Minimal binding / liquid retention Black printed alpha-numerics Virgin, medical-grade polypropylene