Learn More
Alkali Scientific CellPro™ 5ML Serological Pipet, Indiv. Wrap, 400/cs
Catalog No. 502335836
Encompass
Serological Pipets STERILE Individually Wrapped Serological Pipettes color coded paper/plastic wrapping for pop-through or peel-apart techniques Stretched, Non-welded Design Bi-directional and extra Negative Graduation Marks Filter Plug and 100% Polystyrene Gamma Sterilized; Pyrogen/Cytotoxin/RNase & Dnase Free
Catalog No. 502335836 Supplier Alkali Scientific Supplier No. SP205CS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Serological Pipets STERILE Individually Wrapped Serological Pipettes color coded paper/plastic wrapping for pop-through or peel-apart techniques Stretched, Non-welded Design Bi-directional and extra Negative Graduation Marks Filter Plug and 100% Polystyrene Gamma Sterilized; Pyrogen/Cytotoxin/RNase & Dnase Free