Learn More
Alkali Scientific CellPro™ 3.5ml 48-Well Deep Well Plate, U-Bottom, Round Well, 5/pk, 50/cs, DNase, RNase & Pyrogen-Free
Catalog No. 503026733
Encompass
Virgin, heat-resistant, medical-grade polypropylene plates RNase, DNase & pyrogen free Minimal binding & liquid retention Black printed alpha-numeric reference grids U (round) bottoms Dimensions: 5.03" L x 3.36" W x 2.03" Hgt. Autoclavable Temperature range: -86C to 82.2C Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).
Catalog No. 503026733 Supplier Alkali Scientific Supplier No. MPD37
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Virgin, heat-resistant, medical-grade polypropylene plates RNase, DNase & pyrogen free Minimal binding & liquid retention Black printed alpha-numeric reference grids U (round) bottoms Dimensions: 5.03" L x 3.36" W x 2.03" Hgt. Autoclavable Temperature range: -86C to 82.2C Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).