Learn More
Alkali Scientific CellPro 1mL Serological Pipets, Individually Wrapped, Sterile in resealable bulk poly bags 400/pk
Catalog No. 502335846
Encompass
CellPro™ Serological Pipets are sterile and individually wrapped. They have color coded paper/plastic wrapping for pop-through or peel-apart techniques. Designed with quality in mind with a stretched, non-welded design and bi-directional and extra negative graduation marks. They contain a filter plug, are 100% polystyrene, and are Gamma sterilized as well as pyrogen, cytotoxin, RNase and Dnase free.
Catalog No. 502335846 Supplier Alkali Scientific Supplier No. SP201B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CellPro™ Serological Pipets are sterile and individually wrapped. They have color coded paper/plastic wrapping for pop-through or peel-apart techniques. Designed with quality in mind with a stretched, non-welded design and bi-directional and extra negative graduation marks. They contain a filter plug, are 100% polystyrene, and are Gamma sterilized as well as pyrogen, cytotoxin, RNase and Dnase free.