missing translation for 'onlineSavingsMsg'
Learn More

Cellpath EcoSlide Microscope Slide Storage Box

Catalog No. 22222038
Change view
Click to view available options
Device Holds:
1000 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Device Holds
22222038 1000 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22222038 Supplier Cellpath Supplier No. WES010008F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cost effective solution to the long-term storage of microscope slides. CellPath ECOSLIDE Microscope Slide Storage Box is made from recycle material and is supplied with pre-printed archive label for clear identification and rapid retrieval.

  • Supplied assembled or flat-packed
  • Made from recycled material
  • For long term storage of microscope slides
  • Stored in 5 rows separated by full height dividers
  • Includes integral hinged lid
  • Contents can be marked on the end of each box
  • Supplied with pre-printed archive label for clear identification and rapid retrieval

Specifications

Type Microscope Slide Storage
Color Brown
Device Holds 1000 Slides
For Use With (Application) Long-term storage of microscope slides