missing translation for 'onlineSavingsMsg'
Learn More

Cellpath BlocStor 7 Storage Box

Catalog No. 22222039
Change view
Click to view available options
Product Type:
Storage Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22222039 Storage Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22222039 Supplier Cellpath Supplier No. WCB040008F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the long term archiving of tissue blocks. CellPath BlocStor 7 Storage Box is manufactured from recycled fiberboard and is designed for maximum utilisation of storage space.

  • Manufactured from recycled fibreboard
  • Flat-packed for efficient use of space
  • Supplied with a pre-printed archive label for clear identification and rapid retrieval
  • Blocks are stored vertically

Specifications

Product Type Storage Box
Holds Tissue Blocks
For Use With (Application) Tissue Block Storage
Includes Pre-printed Archive Label
Depth (Metric) 17 cm
Height (Metric) 17 cm
Width (Metric) 51 cm