Celloger Stack is an automated multi-layer vessel monitoring system that enables real-time imaging of cell samples while they are cultured in an incubator. The system is capable of detecting cells growing in a multi-layer chamber and identifying the optimal time for cell harvest. With its motorized camera that can move in all directions, the system captures multiple points within the travel range which can be made into a time-lapse video with a simple click. NOTE: WPI is an authorized distributor of