Learn More
Thermo Scientific™ CellInsight™ CX7 High Content Analysis Platform and Store Standard Edition plus Robotic Plate Handling
A fast, automated cellular imaging and analysis platform designed for quantitative microscopy.
Supplier:  Thermo Scientific™ CX7C1115
Encompass
Catalog No. CX7C1115
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
For research use only
Description
Description
Integrated system for high content analysis
The Thermo Scientific™ CellInsight™ CX7 High Content Analysis (HCA) Platform is a fast, automated cellular imaging and analysis platform designed for quantitative microscopy.
The CellInsight CX7 HCA Platform offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes. See how the CellInsight CX7 platform compares to other high content analysis platforms from Thermo Fisher Scientific.
Features of the CellInsight CX7 HCA Platform
  • 7-channel fluorescent imaging
  • 5-channel bright-field imaging
  • 7-channel confocal imaging
  • Software and laser-based autofocus for consistent scan times
  • Configured for fully automated plate handling and scanning
Capabilities
  • Addresses hundreds of assay types with flexible software and hardware tools
  • Scan and analyze microscope slides or most SBS-standard format microplates
  • Use HCS Studio software for integrated instrument control and data analysis
  • Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
  • Compatible with Molecular Probes™ fluorescent detection reagents
Hardware and Software
  • Stable and reliable light source enables reduced variability, increased data quality, and decreased operating costs
  • Broad reagent options with seven-color excitation from violet to near IR for assay multiplexing
  • Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
  • Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
  • HCS Studio Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
  • Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler
Product Suggestions
Product Suggestions