Learn More
Molecular Biology Grade Water Tested to USP Sterile water, Corning™
Catalog No. p-6983003
Change view
Click to view available options
Quantity:
6 x 100 mL
6 x 500 mL
6 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT46000CI 6 x 100 mL
MT46000CV 6 x 500 mL
MT46000CM 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MT46000CI Supplier Corning Incorporated Supplier No. 46000CI
Only null left
Used in RNase, DNase and Protease free applications. Corning™ Molecular Biology Grade Water Tested to USP Sterile water is used for Buffer preparation, DNA/RNA purification, DNA sequencing and libraries, recombinant DNA, cloning, gene mapping.
Molecular biology grade water is ideal for applications where highly pure, RNase-, DNase-, and protease-free water is required (e.g., PCR and other molecular biology reactions setup, DNA/RNA/protein extraction, purification, and storage reagents). Molecular biology grade water is tested to the chemical and physical properties found in the USP monograph for sterile purified water.
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Specifications
Quantity 6 x 100 mL