Learn More
Molecular Biology Grade Water Tested to USP Sterile water, Corning™
Description
Molecular biology grade water is ideal for applications where highly pure, RNase-, DNase-, and protease-free water is required (e.g., PCR and other molecular biology reactions setup, DNA/RNA/protein extraction, purification, and storage reagents). Molecular biology grade water is tested to the chemical and physical properties found in the USP monograph for sterile purified water.
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Specifications
Specifications
| Quantity | 6 x 1 L |