Learn More
Greiner Bio-One CELLCOAT™ Tissue Culture Treated Flask
Used for cell culture applications in most native culture condition.
Supplier: Greiner Bio-One 658950005
Description
- Coated with extracellular matrix proteins or mimetics such as Collagen Type I, Fibronectin, Laminin, Poly-D-Lysine or Poly-L-Lysine
Specifications
| No | |
| 250 mL | |
| Canted | |
| Yes |
| Case | |
| Polystyrene | |
| Collagen Type I Protein | |
| Non-sterile |