Learn More
Greiner Bio-One CELLCOAT™ Tissue Culture Treated Flask
Used for cell culture applications in most native culture condition.
Supplier: Greiner Bio-One 658940005
Description
- Coated with extracellular matrix proteins or mimetics such as Collagen Type I, Fibronectin, Laminin, Poly-D-Lysine or Poly-L-Lysine
Specifications
| No | |
| Case | |
| Polystyrene | |
| Poly-D-Lysine |
| Red | |
| 250 mL | |
| Canted | |
| Non-sterile |