missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ 384-well, Poly-D Lysine-Treated, Flat-Bottom Microplate

Color: Clear; Sterility: Non-sterile; Lid: With Lid
$2,105.14 - $14,419.43
Specifications
| Format | Standard |
|---|---|
| Lid | With Lid |
| Array | 16 x 24 |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Catalog Number | Mfr. No. | Packaging | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Packaging | Price | Quantity & Availability | |||||
07000895
![]() |
Greiner Bio-One
781940 |
5/Pk., 20/Cs. |
Case of 20 Each for $2,105.14
|
|
|||||
07000897
![]() |
Greiner Bio-One
781948 |
20/Pk., 120/Cs. |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Improves adhesion of different cell types to polystyrene surfaces
- Reduced-serum or serum-free cultivation
- Increase in isolation and cultivation efficiency
- Ready-to-use products, time-saving
- Consistent quality
Specifications
| Standard | |
| 16 x 24 | |
| Clear | |
| 384 | |
| Non-sterile | |
| 131 μL | |
| 4800 x g (swinging bucket rotor) |
| With Lid | |
| Not Autoclavable | |
| Polystyrene | |
| 15 to 110 μL | |
| Poly-D-Lysine | |
| Rounded Square | |
| Room Temperature |
Safety and Handling
ShelfLife : 18 Months after month of production