Learn More
Corning™ CellBIND™ 384-Well, CellBIND-Treated, Flat-Bottom Microplate
Description
Corning 384-well microplates for cell culture include tissue culture-treated, Corning CellBIND surface, and Poly-D-Lysine coated microplates.The tissue culture-treated microplates have the same surface treatment used on other Corning cell culture vessels, Features include:
- CellBIND surface treatment provides consistent cell attachment of difficult-to-attach cell lines
- Not a coating; requires no special handling and is stable at room temperature
- Sterile
- Non-Pyrogenic
- Individual alphanumeric codes for well identification
- Opaque walls to prevent well-to-well crosstalk
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Array | 16 x 24 |
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 384 |
| Sterility | Sterile |
| Surface Treatment | CellBIND |
| Well Shape | Flat Bottom (F-well) |