Learn More
Cell Culture Grade Water Tested to USP/EP Sterile Water, Corning™
Catalog No. MT25065LG
Change view
Click to view available options
Quantity:
20 L
100 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT25065LG 100 L
MT25065LB 20 L
MT25065LH 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product should not be used as an injectable or in the preparation of injectables.
Catalog No. MT25065LG Supplier Corning Incorporated Supplier No. 25065LG
Only null left
Product should not be used as an injectable or in the preparation of injectables.
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP/EP Sterile Water are used in preparation of purification and chromatography buffers.
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Specifications
Quantity 100 L