Learn More
Alkali Scientific Cell Culture Chamber Slides, 4 Wells, PS/PP, Glass Slide, Sterile, 12/cs
Catalog No. 502336099
Encompass
ASI chamber slides are designed specifically to cover all your cell-based microscopic research needs and use a patented design that simplifies your work. Our state-of-the-art slides allow you to grow cell cultures within each chamber, making it possible for you to prepare a higher number of samples on a single glass slide. The volume of each well is 2.5mL for the 4 wells cell culture slides. The chambers are produced from high quality clear plastic, and each of the chambers is marked with an alphanumerical code which makes tracking the sample easier. They are sterilized through the use of gamma irradiation, ensuring that this product is ready for use as soon as you receive it. They are also RNase, DNase and Pyrogen free. Materials: Chamber PS / Slide Glass / Holder PP.
Catalog No. 502336099 Supplier Alkali Scientific Supplier No. TCC004
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ASI chamber slides are designed specifically to cover all your cell-based microscopic research needs and use a patented design that simplifies your work. Our state-of-the-art slides allow you to grow cell cultures within each chamber, making it possible for you to prepare a higher number of samples on a single glass slide. The volume of each well is 2.5mL for the 4 wells cell culture slides. The chambers are produced from high quality clear plastic, and each of the chambers is marked with an alphanumerical code which makes tracking the sample easier. They are sterilized through the use of gamma irradiation, ensuring that this product is ready for use as soon as you receive it. They are also RNase, DNase and Pyrogen free. Materials: Chamber PS / Slide Glass / Holder PP.