missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Cell Cryopreservation Medium with 10% DMSO

Catalog No. MP092780248
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP092780248 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP092780248 Supplier MP Biomedicals™ Supplier No. 092780248

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Balanced, protein free, ready to use cryopreservation solution with 10% USP DMSO (CryoPres™).

  • Cell Cryopreservation Medium with 10% DMSO is a balanced, protein free, ready-to-use cryopreservation solution with 10% USP DMSO (Cryopres™)
  • It does not require any dilution or further processing and can be used for cells cultured in serum free medium or with serum containing medium

Specifications

Form Liquid
Sterility Sterile
For Use With (Application) Cryopreservation
Quantity 50 mL
Content And Storage Stored at 2 to 8°C