missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Celite™ R566

Catalog No. 111002463
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002463 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002463 Supplier MilliporeSigma™ Supelco™ Supplier No. 648431KGU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Celite™ R566 is primarily used to interact with target reagents, but it may also be adjusted to fit corresponding scientific methods.

  • Celite™ R566 is an inorganic adsorbent consisting of diatomaceous earth, used for the filtration of analytes from matrices.
  • Material: Silica
  • Grade: Reagent grade
  • Form: Powder
  • Surface Area: 500 m²/g
  • Particle Size: 2000 μm
  • Pore Size: 120 Å mean pore size
  • Separation Technique: Hydrophilic interaction (HILIC)
  • Suitable Application: LPLC

Specifications

Particle Size 20 to 45 μm
Pore Size 120 Å
Technique Chromatography
Grade Reagent Grade
Quantity 1 kg
Surface Area (Metric) 500 m2/g (approx.)
Product Line Celite
Material Silica
Physical Form Powder

Celite is a registered trademark of Imerys Minerals California, Inc.